Gene Transcripts
- Gene Symbol
- HCP5
- Entrez Gene ID
- 10866
- Gene Name
- HLA complex P5 (non-protein coding)
- Gene Aliases
- 6S2650E, D6S2650E, P5-1
- Chromosome Location
- Chr. 6: 31463180 - 31465809 on Build GRCh38
- UniGene ID
- Hs.691948
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000541196.2 |
- |
1, 2 |
105 |
|
Chr. 6 - 31463185 - 31464379 [+] |
GENCODE |
ENST00000460889.5 |
- |
1, 2 |
102 |
|
Chr. 6 - 31463185 - 31463821 [+] |
View Full Product Details