Gene Transcripts
- Gene Symbol
- MCHR2-AS1
- Entrez Gene ID
- 728012
- Gene Name
- MCHR2 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 6: 99994038 - 100076419 on Build GRCh38
- UniGene ID
- Hs.434366
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
HY036177.1 |
- |
3 |
370 |
|
Chr. 6 - 99994079 - 100076384 [+] |
GenBankmRNA |
BU570191.1 |
- |
3 |
336 |
|
Chr. 6 - 99994116 - 100076418 [+] |
GENCODE |
ENST00000430122.6 |
- |
3 |
393 |
|
Chr. 6 - 99994056 - 100076413 [+] |
GenBankmRNA |
BX337371.2 |
- |
3 |
397 |
|
Chr. 6 - 99994052 - 100076406 [+] |
GENCODE |
ENST00000443234.6 |
- |
3 |
302 |
|
Chr. 6 - 99993944 - 100076411 [+] |
View Full Product Details