Gene Transcripts

Gene Symbol
IDI2-AS1
Entrez Gene ID
55853
Gene Name
IDI2 antisense RNA 1
Gene Aliases
C10orf110, HT009, IDI2-AS
Chromosome Location
Chr. 10: 1022637 - 1044201 on Build GRCh38
UniGene ID
Hs.728538
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024628.1 1 48
NR_024629.1 1 48
NR_027708.1 1 48
NR_027709.1 1 48
GenBank mRNA AF220183.1 - 1 19
AK314921.1 - 1 48
BF724542.1 - 11


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CB118196.1 - 1 48 Chr. 10 - 1022637 - 1043579 [+]
GenBankmRNA DA358773.1 - 1 48 Chr. 10 - 1022637 - 1043842 [+]
GENCODE ENST00000420381.5 - 1 17 Chr. 10 - 1022668 - 1043683 [+]
GenBankmRNA AV728065.1 - 1 19 Chr. 10 - 1022666 - 1044105 [+]
GenBankmRNA AV725572.2 - 1 20 Chr. 10 - 1022666 - 1044057 [+]
GenBankmRNA BG034117.1 - 1 25 Chr. 10 - 1022660 - 1043682 [+]
GENCODE ENST00000437374.5 - 1 19 Chr. 10 - 1022666 - 1044198 [+]
GENCODE ENST00000428780.2 - 1 17 Chr. 10 - 1022668 - 1044198 [+]
GenBankmRNA AV728604.1 - 1 19 Chr. 10 - 1022666 - 1043970 [+]
GenBankmRNA BG334515.1 - 1 18 Chr. 10 - 1022667 - 1043900 [+]

View Full Product Details