Gene Transcripts
- Gene Symbol
- FAM13A-AS1
- Entrez Gene ID
- 285512
- Gene Name
- FAM13A antisense RNA 1
- Gene Aliases
- FAM13A1OS, FAM13AOS, NCRNA00039
- Chromosome Location
- Chr. 4: 88709789 - 88730103 on Build GRCh38
- UniGene ID
- Hs.744957
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK093650.1 |
- |
1 |
1134 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000511543.1 |
- |
1 |
1134 |
|
Chr. 4 - 88721715 - 88728686 [+] |
View Full Product Details