Gene Transcripts

Gene Symbol
ROPN1L-AS1
Entrez Gene ID
100505845
Gene Name
ROPN1L antisense RNA 1
Gene Aliases
Chromosome Location
Chr. 5: 10440735 - 10441851 on Build GRCh38
UniGene ID
Hs.599395
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_102746.1 2 236
GenBank mRNA AA393211.1 - 2 89
HY043091.1 - 2 191
HY045761.1 - 2 236


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA HY053860.1 - 2 232 Chr. 5 - 10441058 - 10441847 [-]
GENCODE ENST00000513037.1 - 2 177 Chr. 5 - 10441290 - 10441792 [-]
GenBankmRNA AA398071.1 - 2 174 Chr. 5 - 10441290 - 10441792 [-]

View Full Product Details