Gene Transcripts
- Gene Symbol
- DLGAP2-AS1
- Entrez Gene ID
- 100507435
- Gene Name
- DLGAP2 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 8: 1565509 - 1621664 on Build GRCh38
- UniGene ID
- Hs.667133
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK093412.1 |
- |
1 |
47 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000518009.1 |
- |
1 |
47 |
|
Chr. 8 - 1604552 - 1622417 [-] |
View Full Product Details