Gene Transcripts
- Gene Symbol
- ACKR2
- Entrez Gene ID
- 1238
- Gene Name
- atypical chemokine receptor 2
- Gene Aliases
- CCBP2, CCR10, CCR9, CMKBR9, D6, hD6
- Chromosome Location
- Chr. 3: 42809472 - 42867283 on Build GRCh38
- UniGene ID
- Hs.146346
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC011588.1 |
- |
4 |
718 |
|
BC018716.1 |
- |
4 |
718 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000471537.3 |
- |
4 |
718 |
|
Chr. 3 - 42809472 - 42900336 [+] |
View Full Product Details