Gene Transcripts
- Gene Symbol
- Bcl9
- Entrez Gene ID
- 77578
- Gene Name
- B cell CLL/lymphoma 9
- Gene Aliases
- 2610202E01Rik, 8030475K17Rik, A330041G23Rik, Gm130
- Chromosome Location
- Chr. 3: 97203658 - 97298083 on Build GRCm38
- UniGene ID
- Mm.226175
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK145276.1 |
- |
2 |
1138 |
|
View Full Product Details