Gene Transcripts

Gene Symbol
Rian
Entrez Gene ID
75745
Gene Name
RNA imprinted and accumulated in nucleus
Gene Aliases
5530401N18Rik, C130089L09Rik, Meg8
Chromosome Location
Chr. 12: 109603945 - 109661716 on Build GRCm38
UniGene ID
Mm.293263
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028261.1 5 507
GenBank mRNA AF498299.1 - 2 104
AF498300.1 - 2 104
AF498301.1 - 2 104
EU434919.1 - 5 479


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CB195177.1 - 5 499 Chr. 12 - 109603945 - 109645375 [+]
GenBankmRNA BF152417.1 - 4 222 Chr. 12 - 109604046 - 109627075 [+]
GenBankmRNA AV571224.1 - 3 247 Chr. 12 - 109617222 - 109637887 [+]

View Full Product Details