Gene Transcripts

Gene Symbol
D17H6S53E
Entrez Gene ID
114585
Gene Name
DNA segment, Chr 17, human D6S53E
Gene Aliases
G4, NG34
Chromosome Location
Chr. 17: 35126402 - 35128855 on Build GRCm38
UniGene ID
Mm.42070
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033477.2 NP_258438.1 3 1122
GenBank mRNA BC138707.1 - 1 818
BC138708.1 - 1 818 8861730

View Full Product Details