Gene Transcripts

Gene Symbol
Bhlhe23
Entrez Gene ID
140489
Gene Name
basic helix-loop-helix family, member e23
Gene Aliases
A930001L02Rik, BETA4, Bhlhb4
Chromosome Location
Chr. 2: 180774381 - 180776900 on Build GRCm38
UniGene ID
Mm.134062
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080641.5 NP_542372.2 1 1666
GenBank mRNA AK044215.1 - 1 1666
AK051155.1 - 1 1667

View Full Product Details