Gene Transcripts

Gene Symbol
PAX4
Entrez Gene ID
5078
Gene Name
paired box 4
Gene Aliases
KPD, MODY9
Chromosome Location
Chr. 7: 127610292 - 127618192 on Build GRCh38
UniGene ID
Hs.129706
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006193.2 NP_006184.2 9 1639
GenBank mRNA AF043978.1 - 9 1639

View Full Product Details