Gene Transcripts

Gene Symbol
PCK2
Entrez Gene ID
5106
Gene Name
phosphoenolpyruvate carboxykinase 2, mitochondrial
Gene Aliases
PEPCK, PEPCK-M, PEPCK2
Chromosome Location
Chr. 14: 24094131 - 24104132 on Build GRCh38
UniGene ID
Hs.75812
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018073.2 NP_001018083.2 7 1386
NM_001291556.1 NP_001278485.1 7 1220
NM_001308054.1 NP_001294983.1 7 1647
NM_004563.3 NP_004554.3 7 1386
XM_006720158.2 XP_006720221.1 7 1379
GenBank mRNA AK129934.1 - 7 1204
AK301400.1 - 7 1230
AK307913.1 - 5 1974
AK311710.1 - 5 1703
AK316206.1 - 7 1040
BC001454.2 - 7 1176 3138368
BX248272.1 - 7 1160
CR456913.1 - 7 1111
KU178243.1 - 7 1111
KU178244.1 - 7 1111
X92720.1 - 7 1177

View Full Product Details