Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001161406.1 | NP_001154878.1 | 15 | 1864 | |
NM_145890.2 | NP_665897.2 | 13 | 1496 | ||
GenBank mRNA | AF411211.1 | - | 13 | 1496 | |
AF411212.1 | - | 15 | 1835 | ||
AK076307.1 | - | 15 | 1850 | ||
AK161048.1 | - | 15 | 1746 | ||
BC013080.1 | - | 15 | 1804 | 3499860 | |
BC037233.1 | - | 15 | 1856 | 4951512 |