Gene Transcripts

Gene Symbol
Vps8
Entrez Gene ID
209018
Gene Name
vacuolar protein sorting 8 homolog (S. cerevisiae)
Gene Aliases
AI315068, AU040738, mKIAA0804
Chromosome Location
Chr. 16: 21423118 - 21644681 on Build GRCm38
UniGene ID
Mm.235683
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001285893.1 NP_001272822.1 43 4080
NM_001285894.1 NP_001272823.1 43 3943
GenBank mRNA AK029007.1 - 27 2297
AK031205.1 - 20 2122
AK122375.1 - 43 3892
AK141678.1 - 43 4003
AK219023.1 - 3 261
BC023914.1 - 29 2471 5323126
BC032214.1 - 23 2049 5323693
BC055323.1 - 43 3997 5706288

View Full Product Details