Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173747.3 | NP_776108.2 | 2 | 375 | |
GenBank mRNA | AK028433.1 | - | 2 | 297 | |
AK144167.1 | - | 2 | 305 | ||
BC092224.1 | - | 2 | 281 | 30355916 | |
BC120679.1 | - | 1 | 58 | 40129602 | |
BC125335.1 | - | 1 | 58 | 40129850 | |
CK333382.1 | - | 2 | 350 |