Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020546.2 | NP_065433.2 | 20 | 2561 | |
XM_011513942.2 | XP_011512244.1 | 19 | 2370 | ||
GenBank mRNA | AB028983.1 | - | 17 | 1882 | |
AF070583.1 | - | 4 | 322 | ||
AF410885.1 | - | 20 | 2520 | ||
AK123013.1 | - | 20 | 2408 | ||
AK127036.1 | - | 20 | 2509 | ||
AK294555.1 | - | 17 | 2017 | ||
AK295531.1 | - | 17 | 1901 | ||
AK299775.1 | - | 20 | 1987 | ||
AK304041.1 | - | 20 | 2396 | ||
AK307474.1 | - | 3 | 205 | ||
BC111743.1 | - | 20 | 2499 | 40038765 | |
L21993.1 | - | 7 | 649 | ||
X74210.1 | - | 7 | 649 |