Gene Transcripts

Gene Symbol
PDHB
Entrez Gene ID
5162
Gene Name
pyruvate dehydrogenase (lipoamide) beta
Gene Aliases
PDHBD, PDHE1-B, PHE1B
Chromosome Location
Chr. 3: 58427630 - 58433852 on Build GRCh38
UniGene ID
Hs.161357
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000925.3 NP_000916.2 4 294
NM_001173468.1 NP_001166939.1 4 294
NM_001315536.1 NP_001302465.1 3 240
NR_033384.1 3 364
GenBank mRNA AB209241.1 - 3 1904
AK225569.1 - 4 269
AK225621.1 - 4 274
AK293153.1 - 4 260
AK311514.1 - 4 269
AK313022.1 - 4 269
AL117618.1 - 3 205
BC000439.2 - 4 261 2820672
BC001924.1 - 4 261 2820672
CR541911.1 - 4 251
DA965753.1 - 4 294
DB039833.1 - 3 364
J03576.1 - 4 269
M34055.1 - 4 256
M34056.1 - 3 179
M34479.1 - 4 251
M54788.1 - 4 256
X57778.1 - 4 270

View Full Product Details