Gene Transcripts

Gene Symbol
Kmt2a
Entrez Gene ID
214162
Gene Name
lysine (K)-specific methyltransferase 2A
Gene Aliases
6430520K01, ALL-1, All1, Cxxc7, HRX, HTRX1, Mll, Mll1, mKIAA4050
Chromosome Location
Chr. 9: 44803355 - 44881359 on Build GRCm38
UniGene ID
Mm.2389
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081049.1 NP_001074518.1 16 5105
GenBank mRNA AK220457.1 - 14 2744
L17069.1 - 16 4816

View Full Product Details