Gene Transcripts

Gene Symbol
Taar5
Entrez Gene ID
215854
Gene Name
trace amine-associated receptor 5
Gene Aliases
Gm227, taR-5
Chromosome Location
Chr. 10: 23970706 - 23971719 on Build GRCm38
UniGene ID
Mm.377236
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009574.1 NP_001009574.1 1 326
GenBank mRNA BC139188.1 - 1 371
BC139191.1 - 1 371 8862213

View Full Product Details