Gene Transcripts

Gene Symbol
Tmem5
Entrez Gene ID
216395
Gene Name
transmembrane protein 5
Gene Aliases
6330415D21Rik
Chromosome Location
Chr. 10: 122081120 - 122097366 on Build GRCm38
UniGene ID
Mm.236764
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153059.1 NP_694699.1 6 1034
GenBank mRNA AK043098.1 - 1068
AK075928.1 - 6 1024
AK078061.1 - 6 1102
BC020100.1 - 6 1034 3981114
BC023194.1 - 3 482
BC083125.1 - 7 1202

View Full Product Details