Gene Transcripts

Gene Symbol
Acap1
Entrez Gene ID
216859
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 1
Gene Aliases
Centb1
Chromosome Location
Chr. 11: 69881567 - 69895539 on Build GRCm38
UniGene ID
Mm.288671
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153788.3 NP_722483.2 6, 7 736
GenBank mRNA AK133473.1 - 4, 5 1211
AK138736.1 - 6, 7 728
AK154210.1 - 6, 7 701
AK154449.1 - 6, 7 739
AK155001.1 - 6, 7 757
BC031462.1 - 6, 7 736 4017986
BC037481.1 - 1, 2 363 4460639

View Full Product Details