Gene Transcripts

Gene Symbol
Dhx33
Entrez Gene ID
216877
Gene Name
DEAH (Asp-Glu-Ala-His) box polypeptide 33
Gene Aliases
3110057P17Rik, 9430096J02Rik, Ddx33
Chromosome Location
Chr. 11: 70984091 - 71004553 on Build GRCm38
UniGene ID
Mm.35399
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178367.4 NP_848144.3 7 1196
GenBank mRNA AK035177.1 - 7 1196
BC027842.1 - 7 1193
BC052172.1 - 7 1190 6515901

View Full Product Details