Gene Transcripts

Gene Symbol
Ankrd34b
Entrez Gene ID
218440
Gene Name
ankyrin repeat domain 34B
Gene Aliases
6430502M16Rik, DP58
Chromosome Location
Chr. 13: 92425641 - 92441658 on Build GRCm38
UniGene ID
Mm.213623
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175455.4 NP_780664.2 5 983
GenBank mRNA AK078203.1 - 5 1057
AK090387.1 - 5 986
AK138465.1 - 4 1169
BC096609.1 - 5 859
BC119312.1 - 5 845 8734061
BC119314.1 - 5 845 8734063

View Full Product Details