Gene Transcripts

Gene Symbol
E430025E21Rik
Entrez Gene ID
223593
Gene Name
RIKEN cDNA E430025E21 gene
Gene Aliases
AL022848, C76463, Kiaa0196, mKIAA0196, strumpellin
Chromosome Location
Chr. 15: 59331997 - 59374194 on Build GRCm38
UniGene ID
Mm.218665
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153548.2 NP_705776.2 5 735
GenBank mRNA AK128970.1 - 5 750
AK215294.1 - 4 375
BC067035.1 - 5 735 30532658

View Full Product Details