Gene Transcripts

Gene Symbol
Klhl20
Entrez Gene ID
226541
Gene Name
kelch-like 20
Gene Aliases
AI504637, D930050H05Rik, Kleip, mKIAA4210
Chromosome Location
Chr. 1: 161088375 - 161131508 on Build GRCm38
UniGene ID
Mm.255165
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039482.1 NP_001034571.1 5, 6 1014
GenBank mRNA AK032131.1 - 3, 4 501
AK053102.1 - 5, 6 964
AK054169.1 - 5, 6 982
AK220547.1 - 5, 6 993
BC019571.1 - 5, 6 986 4235202
BC060183.1 - 5, 6 943

View Full Product Details