Gene Transcripts

Gene Symbol
Tstd1
Entrez Gene ID
226654
Gene Name
thiosulfate sulfurtransferase (rhodanese)-like domain containing 1
Gene Aliases
AV065366, EG226654, Gm4848, KAT
Chromosome Location
Chr. 1: 171419033 - 171420352 on Build GRCm38
UniGene ID
Mm.46366
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164525.1 NP_001157997.1 4 500
GenBank mRNA BY720852.1 - 4 469

View Full Product Details