Gene Transcripts

Gene Symbol
Asxl1
Entrez Gene ID
228790
Gene Name
additional sex combs like 1
Gene Aliases
mKIAA0978
Chromosome Location
Chr. 2: 153346139 - 153404007 on Build GRCm38
UniGene ID
Mm.330677
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039939.1 NP_001035028.1 12, 13 1912
GenBank mRNA AK122413.1 - 12, 13 1912

View Full Product Details