Gene Transcripts
- Gene Symbol
- Pogz
- Entrez Gene ID
- 229584
- Gene Name
- pogo transposable element with ZNF domain
- Gene Aliases
- 9530006B08Rik
- Chromosome Location
- Chr. 3: 94823043 - 94883569 on Build GRCm38
- UniGene ID
- Mm.274787
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001165948.1 |
NP_001159420.1
|
3, 4 |
459 |
|
NM_172683.3 |
NP_766271.2
|
3, 4 |
459 |
|
GenBank mRNA |
AK035255.1 |
- |
3, 4 |
314 |
|
AK086062.1 |
- |
3, 4 |
315 |
|
AK161889.1 |
- |
3, 4 |
314 |
|
BC065134.1 |
- |
3, 4 |
448 |
|
BC066108.1 |
- |
3, 4 |
448 |
|
BC085188.1 |
- |
3, 4 |
319 |
30682742 |
BC096492.1 |
- |
2, 3 |
302 |
30745658 |
BC139432.1 |
- |
3, 4 |
314 |
8862454 |
BC139433.1 |
- |
3, 4 |
314 |
8862455 |
CF743949.1 |
- |
3, 4 |
448 |
|
View Full Product Details