Gene Transcripts

Gene Symbol
Foxj3
Entrez Gene ID
230700
Gene Name
forkhead box J3
Gene Aliases
C330039G02Rik, Fhd6
Chromosome Location
Chr. 4: 119539661 - 119629119 on Build GRCm38
UniGene ID
Mm.259227
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172699.3 NP_766287.1 6 780
GenBank mRNA AK082840.1 - 6 781
AK139508.1 - 6 714
BC058231.1 - 6 753 6314981

View Full Product Details