Gene Transcripts

Gene Symbol
Nt5c1a
Entrez Gene ID
230718
Gene Name
5'-nucleotidase, cytosolic IA
Gene Aliases
Cn1a, Gm49
Chromosome Location
Chr. 4: 123201210 - 123225333 on Build GRCm38
UniGene ID
Mm.354635
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085502.1 NP_001078971.1 4 510
GenBank mRNA BC147308.1 - 4 559
BC147309.1 - 4 559 9056378
BC147730.1 - 4 554 9056679
BC147733.1 - 4 554 9056682

View Full Product Details