Gene Transcripts

Gene Symbol
Iqcc
Entrez Gene ID
230767
Gene Name
IQ motif containing C
Gene Aliases
BC026978
Chromosome Location
Chr. 4: 129615617 - 129619213 on Build GRCm38
UniGene ID
Mm.441132
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198026.2 NP_932143.2 5 929
GenBank mRNA BC026978.1 - 3 704
BC031493.1 - 906

View Full Product Details