Gene Transcripts

Gene Symbol
Brat1
Entrez Gene ID
231841
Gene Name
BRCA1-associated ATM activator 1
Gene Aliases
AA881470, Baat1
Chromosome Location
Chr. 5: 140704786 - 140719379 on Build GRCm38
UniGene ID
Mm.271932
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276287.1 NP_001263216.1 2 282
NM_181066.3 NP_851411.1 2 184
GenBank mRNA AK043212.1 - 2 147
AK049321.1 - 2 173
AK084987.1 - 2 173
AK085091.1 - 2 173
AK142387.1 - 2 144
AK154448.1 - 2 155
AK170269.1 - 2 176
AK187162.1 - 1 74
BC068301.1 - 2 248 30061848
BY788789.1 - 2 283

View Full Product Details