Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145589.2 | NP_663564.2 | 7 | 1228 | |
GenBank mRNA | AK133884.1 | - | 7 | 1192 | |
AK139559.1 | - | 8 | 1003 | ||
AK145816.1 | - | 8 | 1008 | ||
AK151203.1 | - | 8 | 1060 | ||
AK188157.1 | - | 1 | 115 | ||
BC055033.1 | - | 7 | 1228 | 6310272 |