Gene Transcripts

Gene Symbol
Slc17a5
Entrez Gene ID
235504
Gene Name
solute carrier family 17 (anion/sugar transporter), member 5
Gene Aliases
4631416G20Rik, 4732491M05, AST, ISSD, NSD, SD, SIALIN, SIASD, SLD
Chromosome Location
Chr. 9: 78536487 - 78588065 on Build GRCm38
UniGene ID
Mm.46932
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172773.3 NP_766361.1 3 455
GenBank mRNA AK029102.1 - 3 438
AK087395.1 - 3 438
AK169868.1 - 3 404

View Full Product Details