Gene Transcripts

Gene Symbol
Adamts14
Entrez Gene ID
237360
Gene Name
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 14
Gene Aliases
Adamts-14, TS14
Chromosome Location
Chr. 10: 61197112 - 61273438 on Build GRCm38
UniGene ID
Mm.333252
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081127.1 NP_001074596.1 13 2182
GenBank mRNA BC157953.1 - 13 2017
BC158014.1 - 13 2015 9088151
BC172106.1 - 13 2012

View Full Product Details