Gene Transcripts

Gene Symbol
Ptprq
Entrez Gene ID
237523
Gene Name
protein tyrosine phosphatase, receptor type, Q
Gene Aliases
Chromosome Location
Chr. 10: 107517360 - 107720078 on Build GRCm38
UniGene ID
Mm.391418
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081432.1 NP_001074901.1 14 2244
GenBank mRNA AK041657.1 - 14 2248

View Full Product Details