Gene Transcripts
- Gene Symbol
- Col23a1
- Entrez Gene ID
- 237759
- Gene Name
- collagen, type XXIII, alpha 1
- Gene Aliases
- 2810458L13Rik, AI429655
- Chromosome Location
- Chr. 11: 51289678 - 51583925 on Build GRCm38
- UniGene ID
- Mm.154093
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_153393.2 |
NP_700442.2
|
29 |
1856 |
|
GenBank mRNA |
AF410792.1 |
- |
29 |
1856 |
|
View Full Product Details