Gene Transcripts

Gene Symbol
Rsad1
Entrez Gene ID
237926
Gene Name
radical S-adenosyl methionine domain containing 1
Gene Aliases
B430319G23, BC056485
Chromosome Location
Chr. 11: 94539798 - 94556861 on Build GRCm38
UniGene ID
Mm.342275
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013381.2 NP_001013399.1 3 553
GenBank mRNA AK046706.1 - 3 558
AK148401.1 - 3 548
BB645699.1 - 3 558
BC139274.1 - 3 496 8862296
BC139276.1 - 3 496 8862298

View Full Product Details