Gene Transcripts

Gene Symbol
MAP2K3
Entrez Gene ID
5606
Gene Name
mitogen-activated protein kinase kinase 3
Gene Aliases
MAPKK3, MEK3, MKK3, PRKMK3, SAPKK-2, SAPKK2
Chromosome Location
Chr. 17: 21284656 - 21315240 on Build GRCh38
UniGene ID
Hs.514012
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316332.1 NP_001303261.1 6 749
NM_002756.4 NP_002747.2 5 366
NM_145109.2 NP_659731.1 5 624
XM_005256723.2 XP_005256780.1 6 664
XM_011523958.2 XP_011522260.1 5 423
XM_011523959.1 XP_011522261.1 7 631
XM_017024857.1 XP_016880346.1 5 493
XM_017024858.1 XP_016880347.1 6 481
XM_017024859.1 XP_016880348.1 7 821
GenBank mRNA AB451226.1 - 5 359
AB451349.1 - 5 359
AK223490.1 - 5 499
AK315505.1 - 5 569
BM836867.1 - 5 604
CR536514.1 - 5 359
CR541793.1 - 5 359
D87116.1 - 5 416
DA987047.1 - 5 367
GQ225578.1 - 5 359
L36719.1 - 7 609
U66839.1 - 5 485

View Full Product Details