Gene Transcripts

Gene Symbol
Olfr38
Entrez Gene ID
258988
Gene Name
olfactory receptor 38
Gene Aliases
18A, MOR257-2
Chromosome Location
Chr. 6: 42762054 - 42763007 on Build GRCm38
UniGene ID
Mm.377777
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146986.1 NP_667197.1 1 637
GenBank mRNA BC106817.1 - 1 636
BC128054.1 - 1 637 40043588

View Full Product Details