Gene Transcripts
- Gene Symbol
- Zkscan17
- Entrez Gene ID
- 268417
- Gene Name
- zinc finger with KRAB and SCAN domains 17
- Gene Aliases
- Nizp1, Zfp496, Znf496
- Chromosome Location
- Chr. 11: 59485520 - 59506640 on Build GRCm38
- UniGene ID
- Mm.239793
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001130529.2 |
NP_001124001.1
|
5, 6 |
1278 |
|
NM_001291014.1 |
NP_001277943.1
|
5, 6 |
1032 |
|
NM_172941.4 |
NP_766529.3
|
5, 6 |
1325 |
|
GenBank mRNA |
AK051711.1 |
- |
5, 6 |
614 |
|
AK139346.1 |
- |
5, 6 |
1568 |
|
AK155196.1 |
- |
5, 6 |
855 |
|
AK155490.1 |
- |
5, 6 |
907 |
|
AK158448.1 |
- |
5, 6 |
863 |
|
AK170924.1 |
- |
5, 6 |
855 |
|
AK170989.1 |
- |
5, 6 |
860 |
|
AK171190.1 |
- |
5, 6 |
907 |
|
AY302242.1 |
- |
4, 5 |
806 |
|
BC040205.1 |
- |
4, 5 |
810 |
4947290 |
CN530086.1 |
- |
3, 4 |
725 |
|
View Full Product Details