Gene Transcripts

Gene Symbol
Fat3
Entrez Gene ID
270120
Gene Name
FAT tumor suppressor homolog 3 (Drosophila)
Gene Aliases
9430076A06Rik, D430038H04Rik, Gm1132, Gm510, MFAT3F
Chromosome Location
Chr. 9: 15910189 - 16502879 on Build GRCm38
UniGene ID
Mm.195010
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080814.1 NP_001074283.1 21 11962
GenBank mRNA AK035028.1 - 4 767
BC117744.1 - 4 742

View Full Product Details