Gene Transcripts
- Gene Symbol
- PTMA
- Entrez Gene ID
- 5757
- Gene Name
- prothymosin, alpha
- Gene Aliases
- TMSA
- Chromosome Location
- Chr. 2: 231708525 - 231713541 on Build GRCh38
- UniGene ID
- Hs.459927
- Species
- Homo sapiens
- Gene Symbol
- MIR1244-1
- Entrez Gene ID
- 100302285
- Gene Name
- microRNA 1244-1
- Gene Aliases
- MIR1244, MIRN1244, hsa-mir-1244, hsa-mir-1244-1
- Chromosome Location
- Chr. 2: 231713314 - 231713398 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_036052.1 |
|
1 |
71 |
|
- Gene Symbol
- MIR1244-2
- Entrez Gene ID
- 100422885
- Gene Name
- microRNA 1244-2
- Gene Aliases
- Chromosome Location
- Chr. 5: 118974586 - 118974670 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_036262.1 |
|
1 |
71 |
|
- Gene Symbol
- MIR1244-3
- Entrez Gene ID
- 100422872
- Gene Name
- microRNA 1244-3
- Gene Aliases
- Chromosome Location
- Chr. 12: 9239467 - 9239551 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_036263.1 |
|
1 |
71 |
|
- Gene Symbol
- MIR1244-4
- Entrez Gene ID
- 103504734
- Gene Name
- microRNA 1244-4
- Gene Aliases
- hsa-mir-1244-4
- Chromosome Location
- Chr. 12: 12111952 - 12112036 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_128710.1 |
|
1 |
71 |
|
View Full Product Details