Gene Transcripts
- Gene Symbol
- Rfx7
- Entrez Gene ID
- 319758
- Gene Name
- regulatory factor X, 7
- Gene Aliases
- 2510005N23Rik, 9930116O05Rik, D130086K05Rik, Rfxdc2
- Chromosome Location
- Chr. 9: 72532167 - 72622949 on Build GRCm38
- UniGene ID
- Mm.32081
- Species
- Mus musculus
View Full Product Details