Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177016.3 | NP_795990.2 | 8 | 1048 | |
GenBank mRNA | AK033669.1 | - | 8 | 1051 | |
AK050272.1 | - | 7 | 1162 | ||
AK162320.1 | - | 7 | 1491 | ||
BC147415.1 | - | 8 | 1009 | 9056491 | |
BC147416.1 | - | 8 | 1009 | 9056498 |