Gene Transcripts
- Gene Symbol
- Trdv2-2
- Entrez Gene ID
- 320913
- Gene Name
- T cell receptor delta variable 2-2
- Gene Aliases
- Gm17015
- Chromosome Location
- Chr. 14: 53960993 - 53961602 on Build GRCm38
- UniGene ID
- Mm.303179
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AA756275.1 |
- |
2 |
366 |
|
View Full Product Details