Gene Transcripts

Gene Symbol
Sh2d4b
Entrez Gene ID
328381
Gene Name
SH2 domain containing 4B
Gene Aliases
A430109M18Rik, D030001E08
Chromosome Location
Chr. 14: 40811521 - 40902337 on Build GRCm38
UniGene ID
Mm.235694
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177816.3 NP_808484.2 5 803
GenBank mRNA AK050671.1 - 2 517
AK134033.1 - 2 293
AX776199.1 - 5 803
BC138964.1 - 5 901 8861986

View Full Product Details