Gene Transcripts

Gene Symbol
Skint9
Entrez Gene ID
329918
Gene Name
selection and upkeep of intraepithelial T cells 9
Gene Aliases
A030013N09Rik, Skint-9
Chromosome Location
Chr. 4: 112385969 - 112433985 on Build GRCm38
UniGene ID
Mm.246783
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177864.2 NP_808532.1 5 642
GenBank mRNA AK037244.1 - 5 642
EF494907.1 - 5 634

View Full Product Details