Gene Transcripts

Gene Symbol
Ksr2
Entrez Gene ID
333050
Gene Name
kinase suppressor of ras 2
Gene Aliases
Chromosome Location
Chr. 5: 117413770 - 117775004 on Build GRCm38
UniGene ID
Mm.437947
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114545.2 NP_001108017.2 4 1512
GenBank mRNA AK137433.1 - 4 1514
BC068233.1 - 2 297
BC150883.1 - 4 1512
BC150890.1 - 4 1512
DQ531035.1 - 4 685

View Full Product Details